Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C12orf10
Entrez Gene ID
60314
Gene Name
chromosome 12 open reading frame 10
Gene Aliases
Gamm1, MST024, MSTP024, MYG, MYG1
Chromosome Location
Chr.12: 53299686 - 53307181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021640.3 NP_067653.3 4 - 5 696 80
GenBank mRNA AF111805.1 - 2 - 3 288 80
AF289485.1 - 4 - 5 664 80
BC028904.1 - 3 - 4 600 80
BC051871.1 - 4 - 5 655 80

View Full Product Details