The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000818.2 | NP_000809.1 | 4 - 5 | 1020 | 101 |
NM_001134366.1 | NP_001127838.1 | 4 - 5 | 1020 | 101 | |
GenBank mRNA | BC126327.1 | - | 4 - 5 | 542 | 101 |
BC126329.1 | - | 4 - 5 | 542 | 101 | |
M74826.1 | - | 4 - 5 | 658 | 101 | |
M81882.1 | - | 4 - 5 | 569 | 101 | |
X69936.1 | - | 4 - 5 | 562 | 101 |