The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006660.4 | NP_006651.2 | 4 - 5 | 779 | 95 |
NR_133680.1 | 4 - 5 | 779 | 95 | ||
XM_011521164.2 | XP_011519466.1 | 4 - 5 | 763 | 95 | |
GenBank mRNA | AJ006267.3 | - | 4 - 5 | 584 | 95 |
AK292316.1 | - | 4 - 5 | 705 | 95 | |
AK308133.1 | - | 4 - 5 | 699 | 95 | |
AK310614.1 | - | 4 - 5 | 701 | 95 | |
AL136922.1 | - | 4 - 5 | 682 | 95 | |
BC130373.1 | - | 4 - 5 | 575 | 95 | |
BC136487.1 | - | 4 - 5 | 575 | 95 |