Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BIRC6
Entrez Gene ID
57448
Gene Name
baculoviral IAP repeat containing 6
Gene Aliases
APOLLON, BRUCE
Chromosome Location
Chr.2: 32356907 - 32618899 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016252.3 NP_057336.3 24 - 25 5077 64
XM_005264449.4 XP_005264506.2 24 - 25 5252 64
XM_005264450.4 XP_005264507.2 24 - 25 5240 64
XM_005264451.4 XP_005264508.2 24 - 25 5252 64
XM_005264452.4 XP_005264509.2 24 - 25 4955 64
XM_005264453.4 XP_005264510.2 24 - 25 4955 64
XM_005264454.4 XP_005264511.2 24 - 25 5252 64
XM_005264455.4 XP_005264512.2 23 - 24 5120 64
XM_006712054.3 XP_006712117.1 24 - 25 5252 64
XM_006712055.3 XP_006712118.1 24 - 25 5252 64
XM_011533003.2 XP_011531305.1 24 - 25 4697 64
XM_011533005.2 XP_011531307.1 24 - 25 5252 64
XM_017004556.1 XP_016860045.1 24 - 25 5252 64
XM_017004559.1 XP_016860048.1 24 - 25 5252 64
XM_017004560.1 XP_016860049.1 24 - 25 5252 64
GenBank mRNA AF265555.1 - 24 - 25 4859 64

View Full Product Details