The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016252.3 | NP_057336.3 | 34 - 35 | 6985 | 79 |
XM_005264449.4 | XP_005264506.2 | 34 - 35 | 7187 | 79 | |
XM_005264450.4 | XP_005264507.2 | 34 - 35 | 7175 | 79 | |
XM_005264451.4 | XP_005264508.2 | 34 - 35 | 7160 | 79 | |
XM_005264452.4 | XP_005264509.2 | 34 - 35 | 6890 | 79 | |
XM_005264453.4 | XP_005264510.2 | 34 - 35 | 6863 | 79 | |
XM_005264454.4 | XP_005264511.2 | 34 - 35 | 7187 | 79 | |
XM_005264455.4 | XP_005264512.2 | 33 - 34 | 7055 | 79 | |
XM_006712054.3 | XP_006712117.1 | 34 - 35 | 7169 | 79 | |
XM_006712055.3 | XP_006712118.1 | 34 - 35 | 7187 | 79 | |
XM_006712056.3 | XP_006712119.1 | 34 - 35 | 7088 | 79 | |
XM_011533003.2 | XP_011531305.1 | 34 - 35 | 6632 | 79 | |
XM_011533005.2 | XP_011531307.1 | 34 - 35 | 7187 | 79 | |
XM_017004556.1 | XP_016860045.1 | 34 - 35 | 7169 | 79 | |
XM_017004557.1 | XP_016860046.1 | 34 - 35 | 7076 | 79 | |
XM_017004558.1 | XP_016860047.1 | 34 - 35 | 7046 | 79 | |
XM_017004559.1 | XP_016860048.1 | 34 - 35 | 7187 | 79 | |
XM_017004560.1 | XP_016860049.1 | 34 - 35 | 7169 | 79 | |
GenBank mRNA | AB033115.2 | - | 5 - 6 | 665 | 79 |
AF265555.1 | - | 34 - 35 | 6767 | 79 |