Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BIRC6
Entrez Gene ID
57448
Gene Name
baculoviral IAP repeat containing 6
Gene Aliases
APOLLON, BRUCE
Chromosome Location
Chr.2: 32356907 - 32618899 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016252.3 NP_057336.3 43 - 44 8343 151
XM_005264449.4 XP_005264506.2 43 - 44 8545 151
XM_005264450.4 XP_005264507.2 43 - 44 8533 151
XM_005264451.4 XP_005264508.2 43 - 44 8518 151
XM_005264452.4 XP_005264509.2 43 - 44 8248 151
XM_005264453.4 XP_005264510.2 43 - 44 8221 151
XM_005264454.4 XP_005264511.2 43 - 44 8545 151
XM_005264455.4 XP_005264512.2 42 - 43 8413 151
XM_006712054.3 XP_006712117.1 43 - 44 8527 151
XM_006712055.3 XP_006712118.1 43 - 44 8545 151
XM_006712056.3 XP_006712119.1 43 - 44 8446 151
XM_011533003.2 XP_011531305.1 43 - 44 7990 151
XM_011533005.2 XP_011531307.1 43 - 44 8545 151
XM_017004556.1 XP_016860045.1 43 - 44 8527 151
XM_017004557.1 XP_016860046.1 43 - 44 8434 151
XM_017004558.1 XP_016860047.1 43 - 44 8404 151
XM_017004559.1 XP_016860048.1 43 - 44 8545 151
XM_017004560.1 XP_016860049.1 43 - 44 8527 151
GenBank mRNA AB033115.2 - 14 - 15 2023 151
AF265555.1 - 43 - 44 8125 151

View Full Product Details