Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARFIP2
Entrez Gene ID
23647
Gene Name
ADP ribosylation factor interacting protein 2
Gene Aliases
POR1
Chromosome Location
Chr.11: 6474683 - 6481479 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242854.1 NP_001229783.1 4 - 5 712 91
NM_001242855.1 NP_001229784.1 3 - 4 472 91
NM_012402.3 NP_036534.1 4 - 5 573 91
XM_005252840.4 XP_005252897.1 4 - 5 477 91
GenBank mRNA AK026390.1 - 4 - 5 405 91
AK301856.1 - 3 - 4 427 91
AK301972.1 - 4 - 5 514 91
AK311086.1 - 3 - 4 1055 91
BC000392.2 - 4 - 5 395 91
BM928413.1 - - 540 91
U52522.1 - 4 - 5 389 91
X97567.1 - 2 - 3 208 91

View Full Product Details