The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001973.2 | NP_001001973.1 | 6 - 7 | 741 | 93 |
NM_001320886.1 | NP_001307815.1 | 5 - 6 | 706 | 93 | |
NM_005174.3 | NP_005165.1 | 6 - 7 | 741 | 93 | |
XM_017016290.1 | XP_016871779.1 | 5 - 6 | 651 | 93 | |
GenBank mRNA | AK292896.1 | - | 6 - 7 | 668 | 93 |
AK296803.1 | - | 5 - 6 | 633 | 93 | |
BC000470.2 | - | 6 - 7 | 656 | 93 | |
BC000931.3 | - | 6 - 7 | 689 | 93 | |
BC013394.2 | - | 6 - 7 | 656 | 93 | |
BC013865.1 | - | 6 - 7 | 654 | 93 | |
BC016812.1 | - | 6 - 7 | 671 | 93 | |
BC020824.1 | - | 6 - 7 | 677 | 93 | |
BC026049.1 | - | 6 - 7 | 616 | 93 | |
BQ893894.1 | - | 6 - 7 | 716 | 93 | |
CR457403.1 | - | 6 - 7 | 637 | 93 | |
D16562.1 | - | 6 - 7 | 668 | 93 | |
D16563.1 | - | 6 - 7 | 668 | 93 |