The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301683.1 | NP_001288612.1 | 15 - 16 | 2142 | 76 |
NM_001323584.1 | NP_001310513.1 | 14 - 15 | 2055 | 76 | |
NM_001323585.1 | NP_001310514.1 | 16 - 17 | 2215 | 76 | |
NM_001323586.1 | NP_001310515.1 | 15 - 16 | 2080 | 76 | |
NM_001323587.1 | NP_001310516.1 | 13 - 14 | 1929 | 76 | |
NM_001323588.1 | NP_001310517.1 | 13 - 14 | 1977 | 76 | |
NM_003171.4 | NP_003162.2 | 14 - 15 | 2007 | 76 | |
NR_136626.1 | 14 - 15 | 1931 | 76 | ||
NR_136627.1 | 15 - 16 | 2066 | 76 | ||
GenBank mRNA | AF042169.1 | - | 14 - 15 | 1919 | 76 |
AK290416.1 | - | 14 - 15 | 1961 | 76 | |
AK299670.1 | - | 15 - 16 | 2096 | 76 | |
BC036112.1 | - | 14 - 15 | 1974 | 76 | |
BM675613.1 | - | 1 - 2 | 76 |