The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042603.2 | NP_001036068.1 | 13 - 14 | 2138 | 86 |
GenBank mRNA | AB209999.1 | - | 13 - 14 | 1984 | 86 |
AK225060.1 | - | 13 - 14 | 1920 | 86 | |
AK301150.1 | - | 12 - 13 | 1825 | 86 | |
AK301273.1 | - | 4 - 5 | 820 | 86 | |
AK307580.1 | - | 13 - 14 | 1937 | 86 | |
AK307689.1 | - | 6 - 7 | 743 | 86 | |
BC048307.1 | - | 13 - 14 | 2123 | 86 | |
BC053893.1 | - | 13 - 14 | 2136 | 86 | |
BC110916.1 | - | 13 - 14 | 1924 | 86 | |
S66431.1 | - | 13 - 14 | 1926 | 86 |