The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042603.2 | NP_001036068.1 | 3 - 4 | 731 | 90 |
GenBank mRNA | AB209999.1 | - | 3 - 4 | 577 | 90 |
AK225060.1 | - | 3 - 4 | 513 | 90 | |
AK307580.1 | - | 3 - 4 | 530 | 90 | |
BC048307.1 | - | 3 - 4 | 716 | 90 | |
BC053893.1 | - | 3 - 4 | 729 | 90 | |
BC110916.1 | - | 3 - 4 | 517 | 90 | |
DB079061.1 | - | 3 - 4 | 421 | 90 | |
S66431.1 | - | 3 - 4 | 519 | 90 |