Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATF7IP2
Entrez Gene ID
80063
Gene Name
activating transcription factor 7 interacting protein 2
Gene Aliases
MCAF2
Chromosome Location
Chr.16: 10386055 - 10483640 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256160.1 NP_001243089.1 8 - 9 1615 129
NM_024997.3 NP_079273.2 7 - 8 1572 129
XM_006720953.3 XP_006721016.1 6 - 7 1399 129
XM_011522666.2 XP_011520968.1 3 - 4 335 129
XM_017023705.1 XP_016879194.1 6 - 7 2540 129
XM_017023706.1 XP_016879195.1 6 - 7 2540 129
XM_017023708.1 XP_016879197.1 5 - 6 623 129
XM_017023709.1 XP_016879198.1 2 - 3 934 129
XM_017023710.1 XP_016879199.1 6 - 7 1224 129
GenBank mRNA AK022730.1 - 3 - 4 332 129
AK093414.1 - 8 - 9 1613 129
AK223492.1 - 3 - 4 383 129
AY560615.1 - 5 - 6 1342 129
BC033891.1 - 7 - 8 1568 129
BC069695.1 - 3 - 4 309 129
BC069713.1 - 3 - 4 309 129
BC069730.1 - 3 - 4 309 129
BC137079.1 - 6 - 7 1471 129

View Full Product Details