The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020240.2 | NP_064625.1 | 2 - 3 | 327 | 75 |
XM_017009648.1 | XP_016865137.1 | 2 - 3 | 288 | 75 | |
GenBank mRNA | BC050430.1 | - | 2 - 3 | 261 | 75 |
BC058909.1 | - | 2 - 3 | 236 | 75 | |
BI460220.1 | - | 2 - 3 | 352 | 75 | |
BI522591.1 | - | 2 - 3 | 327 | 75 |