Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC6A13
Entrez Gene ID
6540
Gene Name
solute carrier family 6 member 13
Gene Aliases
GAT-2, GAT2, GAT3
Chromosome Location
Chr.12: 220621 - 262930 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190997.2 NP_001177926.1 12 - 13 1499 67
NM_016615.4 NP_057699.2 14 - 15 1775 67
XM_006719008.3 XP_006719071.1 9 - 10 1562 67
XM_011521012.2 XP_011519314.1 14 - 15 1843 67
XM_011521013.1 XP_011519315.1 15 - 16 1911 67
XM_011521014.1 XP_011519316.1 13 - 14 1667 67
XM_017019842.1 XP_016875331.1 10 - 11 1660 67
XM_017019843.1 XP_016875332.1 10 - 11 1258 67
XM_017019845.1 XP_016875334.1 10 - 11 1160 67
GenBank mRNA AK296127.1 - 12 - 13 1462 67
AK296209.1 - 15 - 16 1706 67
AK313511.1 - 14 - 15 1731 67
BC022392.1 - 14 - 15 1738 67
U76343.2 - 14 - 15 1619 67

View Full Product Details