The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001310156.1 | NP_001297085.1 | 10 - 11 | 1204 | 106 |
GenBank mRNA | AF306347.1 | - | 10 - 11 | 1172 | 106 |
BC119641.1 | - | 10 - 11 | 1108 | 106 | |
BC119642.1 | - | 10 - 11 | 1108 | 106 | |
BC127634.1 | - | 10 - 11 | 1104 | 106 |