The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001310156.1 | NP_001297085.1 | 8 - 9 | 970 | 145 |
XM_011542514.2 | XP_011540816.1 | 8 - 9 | 970 | 145 | |
XM_011542516.1 | XP_011540818.1 | 8 - 9 | 966 | 141 | |
XM_011542517.1 | XP_011540819.1 | 8 - 9 | 966 | 141 | |
GenBank mRNA | BC119641.1 | - | 8 - 9 | 874 | 145 |
BC119642.1 | - | 8 - 9 | 874 | 145 | |
BC127634.1 | - | 8 - 9 | 870 | 141 |