Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIST1H2AK
Entrez Gene ID
8330
Gene Name
histone cluster 1, H2ak
Gene Aliases
H2A/d, H2AFD
Chromosome Location
Chr.6: 27837880 - 27838339 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC034487.2 - 1 - 1 789 88

View Full Product Details