The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002661.4 | NP_002652.2 | 15 - 16 | 1679 | 133 |
GenBank mRNA | AB208914.1 | - | 15 - 16 | 1635 | 133 |
AK304722.1 | - | 12 - 13 | 1083 | 133 | |
AK310042.1 | - | 14 - 15 | 1709 | 133 | |
AY364256.1 | - | 14 - 15 | 1465 | 133 | |
BC007565.1 | - | 15 - 16 | 1612 | 133 | |
BC011772.2 | - | 15 - 16 | 1612 | 133 | |
BC014561.2 | - | 15 - 16 | 1612 | 133 | |
BC018646.2 | - | 15 - 16 | 1612 | 133 | |
M37238.1 | - | 15 - 16 | 1617 | 133 | |
X14034.1 | - | 15 - 16 | 1617 | 133 |