The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002264.3 | NP_002255.3 | 5 - 6 | 623 | 92 |
XM_005247437.3 | XP_005247494.1 | 5 - 6 | 505 | 92 | |
XM_011512795.2 | XP_011511097.1 | 6 - 7 | 557 | 92 | |
GenBank mRNA | BC002374.2 | - | 5 - 6 | 612 | 92 |
BC003009.1 | - | 5 - 6 | 612 | 92 | |
BC090864.1 | - | 5 - 6 | 594 | 92 | |
BT006959.1 | - | 4 - 5 | 435 | 92 | |
CR456743.1 | - | 4 - 5 | 435 | 92 | |
S75295.1 | - | 5 - 6 | 92 | ||
U20620.1 | - | 3 - 4 | 288 | 92 |