The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020805.2 | NP_065856.1 | 2 - 3 | 1338 | 91 |
XM_011526109.2 | XP_011524411.2 | 2 - 3 | 1030 | 91 | |
GenBank mRNA | AB037805.1 | - | 2 - 3 | 1338 | 91 |
AK294996.1 | - | 1 - 2 | 576 | 91 | |
BC021267.2 | - | 2 - 3 | 1206 | 91 |