Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL14
Entrez Gene ID
57565
Gene Name
kelch like family member 14
Gene Aliases
-
Chromosome Location
Chr.18: 32672673 - 32773011 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020805.2 NP_065856.1 3 - 4 1456 111
GenBank mRNA AB037805.1 - 3 - 4 1456 111
AK294996.1 - 2 - 3 694 111

View Full Product Details