The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278425.1 | NP_001265354.1 | 19 - 20 | 3282 | 67 |
NM_052892.3 | NP_443124.3 | 30 - 31 | 5188 | 67 | |
NR_126532.1 | 30 - 31 | 5188 | 67 | ||
GenBank mRNA | AB067466.1 | - | 21 - 22 | 4601 | 67 |
AY164483.1 | - | 30 - 31 | 5188 | 67 | |
AY371495.1 | - | 19 - 20 | 3244 | 67 |