The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004616.2 | NP_004607.1 | 7 - 8 | 750 | 160 |
XM_006719583.3 | XP_006719646.1 | 7 - 8 | 704 | 160 | |
XM_017019913.1 | XP_016875402.1 | 6 - 7 | 786 | 160 | |
GenBank mRNA | AK223022.1 | - | 6 - 7 | 678 | 160 |
AK313112.1 | - | 6 - 7 | 733 | 160 | |
BC005246.1 | - | 7 - 8 | 750 | 160 | |
BC070168.1 | - | 11 - 12 | 1124 | 160 | |
M35252.1 | - | 6 - 7 | 708 | 160 |