Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPA14
Entrez Gene ID
51182
Gene Name
heat shock protein family A (Hsp70) member 14
Gene Aliases
HSP70-4, HSP70L1
Chromosome Location
Chr.10: 14838160 - 14871741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278205.1 NP_001265134.1 1 - 2 299 94
NM_016299.3 NP_057383.2 1 - 2 299 94
NR_103464.1 1 - 2 299 94
GenBank mRNA AF112210.1 - 1 - 2 118 94
AK000880.1 - 1 - 2 140 94
AK225265.1 - 1 - 2 143 94
AK225651.1 - 1 - 2 143 94
AK292323.1 - 1 - 2 107 94
AK302452.1 - 1 - 2 143 94
BC065281.1 - 1 - 2 130 94
BC108738.1 - 1 - 2 233 94
BC119690.1 - 1 - 2 118 94
BP201211.1 - 1 - 2 299 94
BQ424016.1 - 1 - 2 230 94
CN308284.1 - 1 - 2 136 94

View Full Product Details