The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181575.4 | NP_853553.1 | 2 - 3 | 457 | 85 |
NR_126510.1 | 2 - 3 | 457 | 85 | ||
NR_126511.1 | 2 - 3 | 457 | 85 | ||
GenBank mRNA | AF100746.1 | - | 2 - 3 | 230 | 85 |
AF100753.1 | - | 2 - 3 | 257 | 85 | |
AF100754.1 | - | 2 - 3 | 257 | 85 | |
AF165515.1 | - | 2 - 3 | 237 | 85 | |
AK023983.1 | - | 2 - 3 | 235 | 85 | |
AK293121.1 | - | 2 - 3 | 276 | 85 | |
AK301398.1 | - | 2 - 3 | 256 | 85 | |
AK308231.1 | - | 2 - 3 | 258 | 85 | |
AK310159.1 | - | 2 - 3 | 258 | 85 | |
BC001658.1 | - | 2 - 3 | 320 | 85 | |
BC033646.2 | - | 2 - 3 | 272 | 85 | |
CR999537.1 | - | 2 - 3 | 443 | 85 | |
CX164419.1 | - | 2 - 3 | 239 | 85 |