The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001975.2 | NP_001966.1 | 8 - 9 | 1089 | 74 |
GenBank mRNA | AK124656.1 | - | 7 - 8 | 1493 | 74 |
AK290525.1 | - | 8 - 9 | 1215 | 74 | |
AK295220.1 | - | 6 - 7 | 821 | 74 | |
AK312402.1 | - | 8 - 9 | 956 | 74 | |
BC002745.2 | - | 8 - 9 | 943 | 74 | |
BT007383.1 | - | 7 - 8 | 867 | 74 | |
CR536582.1 | - | 7 - 8 | 867 | 74 | |
EU794607.1 | - | 7 - 8 | 867 | 74 | |
M22349.1 | - | 8 - 9 | 941 | 74 | |
M36768.1 | - | 6 - 7 | 789 | 74 | |
X13120.1 | - | 7 - 8 | 864 | 74 | |
X14327.1 | - | 7 - 8 | 864 | 74 |