The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297602.1 | NP_001284531.1 | 5 - 6 | 1479 | 82 |
NM_031299.5 | NP_112589.1 | 5 - 6 | 1554 | 82 | |
NR_123727.1 | 5 - 6 | 1538 | 82 | ||
GenBank mRNA | AK291421.1 | - | 5 - 6 | 757 | 82 |
AK312843.1 | - | 5 - 6 | 774 | 82 | |
BC002551.2 | - | 5 - 6 | 749 | 82 | |
BM911472.1 | - | 5 - 6 | 664 | 82 |