Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WBP2NL
Entrez Gene ID
164684
Gene Name
WBP2 N-terminal like
Gene Aliases
GRAMD7, PAWP
Chromosome Location
Chr.22: 41998725 - 42058456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152613.2 NP_689826.2 1 - 2 149 55
GenBank mRNA AK128138.1 - 1 - 2 149 55
AK129656.1 - 1 - 2 102 55
BC022546.1 - 1 - 2 86 55
BC022549.1 - 1 - 2 86 55
CR456400.1 - 1 - 2 62 55
DB079211.1 - 1 - 2 149 55

View Full Product Details