The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016937.3 | NP_058633.2 | 18 - 19 | 1919 | 109 |
XM_005274552.3 | XP_005274609.1 | 18 - 19 | 2051 | 109 | |
XM_006724499.2 | XP_006724562.1 | 17 - 18 | 1861 | 109 | |
XM_011545540.2 | XP_011543842.1 | 18 - 19 | 2006 | 109 | |
XM_011545541.2 | XP_011543843.1 | 18 - 19 | 2007 | 109 | |
XM_017029594.1 | XP_016885083.1 | 18 - 19 | 2007 | 109 | |
XM_017029595.1 | XP_016885084.1 | 18 - 19 | 2007 | 109 | |
XM_017029596.1 | XP_016885085.1 | 18 - 19 | 2007 | 109 | |
GenBank mRNA | X06745.1 | - | 18 - 19 | 1920 | 109 |