The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100159.2 | NP_001093629.1 | 3 - 4 | 465 | 115 |
NM_001267865.1 | NP_001254794.1 | 2 - 3 | 309 | 115 | |
NM_001267866.1 | NP_001254795.1 | 2 - 3 | 309 | 115 | |
XM_006715649.3 | XP_006715712.1 | 2 - 3 | 325 | 115 | |
XM_006715650.2 | XP_006715713.1 | 2 - 3 | 292 | 115 | |
XM_011515121.1 | XP_011513423.1 | 3 - 4 | 369 | 115 | |
XM_011515122.2 | XP_011513424.1 | 2 - 3 | 325 | 115 | |
XM_011515123.2 | XP_011513425.1 | 2 - 3 | 325 | 115 | |
GenBank mRNA | BC031107.1 | - | 3 - 4 | 456 | 115 |
BC045737.1 | - | 2 - 3 | 309 | 115 | |
BI560654.1 | - | 2 - 3 | 305 | 115 |