The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017631.5 | NP_060101.3 | 18 - 19 | 2862 | 65 |
XM_011532103.2 | XP_011530405.1 | 18 - 19 | 2862 | 65 | |
XM_011532104.2 | XP_011530406.1 | 18 - 19 | 2862 | 65 | |
XM_017008383.1 | XP_016863872.1 | 18 - 19 | 2862 | 65 | |
XM_017008384.1 | XP_016863873.1 | 18 - 19 | 2862 | 65 | |
GenBank mRNA | AK001649.1 | - | 18 - 19 | 2738 | 65 |
AK307404.1 | - | 18 - 19 | 2862 | 65 | |
BC007820.2 | - | 3 - 4 | 389 | 65 | |
BC038115.2 | - | 18 - 19 | 2807 | 65 |