Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX60
Entrez Gene ID
55601
Gene Name
DEXD/H-box helicase 60
Gene Aliases
-
Chromosome Location
Chr.4: 168216291 - 168318807 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017631.5 NP_060101.3 4 - 5 556 140
XM_011532103.2 XP_011530405.1 4 - 5 556 140
XM_011532104.2 XP_011530406.1 4 - 5 556 140
XM_017008383.1 XP_016863872.1 4 - 5 556 140
XM_017008384.1 XP_016863873.1 4 - 5 556 140
GenBank mRNA AK001649.1 - 4 - 5 432 140
AK307404.1 - 4 - 5 556 140
BC038115.2 - 4 - 5 501 140

View Full Product Details