Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPATA18
Entrez Gene ID
132671
Gene Name
spermatogenesis associated 18
Gene Aliases
Mieap, SPETEX1
Chromosome Location
Chr.4: 52051331 - 52097305 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297608.1 NP_001284537.1 2 - 3 561 75
NM_145263.3 NP_660306.1 2 - 3 561 75
NR_123728.1 2 - 3 561 75
XM_005265716.1 XP_005265773.1 2 - 3 476 75
XM_005265717.2 XP_005265774.1 2 - 3 476 75
XM_011534369.1 XP_011532671.1 2 - 3 476 75
XM_011534370.1 XP_011532672.1 2 - 3 476 75
GenBank mRNA AB465501.1 - 2 - 3 187 75
AB465502.1 - 2 - 3 187 75
AK057468.1 - 2 - 3 473 75
AK304386.1 - 2 - 3 476 75
AK314799.1 - 2 - 3 431 75
BC025396.1 - 2 - 3 468 75

View Full Product Details