The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174937.3 | NP_777597.2 | 9 - 10 | 1482 | 96 |
XM_011539577.1 | XP_011537879.1 | 10 - 11 | 1515 | 96 | |
GenBank mRNA | AK096269.1 | - | 9 - 10 | 3259 | 96 |
BC042951.1 | - | 9 - 10 | 1487 | 96 | |
BC093639.1 | - | 9 - 10 | 1331 | 96 | |
BC101535.1 | - | 9 - 10 | 1331 | 96 |