The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161587.1 | NP_001155059.1 | 10 - 11 | 1467 | 75 |
NM_002103.4 | NP_002094.2 | 11 - 12 | 1659 | 75 | |
NR_027763.1 | 10 - 11 | 1477 | 75 | ||
GenBank mRNA | AK223579.1 | - | 11 - 12 | 1615 | 75 |
AK291672.1 | - | 11 - 12 | 1615 | 75 | |
AK294349.1 | - | 6 - 7 | 575 | 75 | |
AK300369.1 | - | 11 - 12 | 1447 | 75 | |
AK302738.1 | - | 10 - 11 | 1433 | 75 | |
BC002617.2 | - | 11 - 12 | 1598 | 75 | |
BC003182.1 | - | 10 - 11 | 1424 | 75 | |
BC007688.2 | - | 11 - 12 | 1585 | 75 | |
J04501.1 | - | 11 - 12 | 1578 | 75 | |
U32573.1 | - | 11 - 12 | 1557 | 75 |