The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190918.1 | NP_001177847.1 | 6 - 7 | 1154 | 64 |
NM_001190919.1 | NP_001177848.1 | 6 - 7 | 1135 | 64 | |
NM_003250.5 | NP_003241.2 | 6 - 7 | 1154 | 64 | |
NM_199334.3 | NP_955366.1 | 6 - 7 | 1154 | 64 | |
GenBank mRNA | AB209346.1 | - | 6 - 7 | 1160 | 64 |
AB307686.1 | - | 5 - 6 | 579 | 64 | |
AK290071.1 | - | 6 - 7 | 981 | 64 | |
AK290530.1 | - | 6 - 7 | 1135 | 64 | |
AK311344.1 | - | 6 - 7 | 929 | 64 | |
BC000261.1 | - | 6 - 7 | 1003 | 64 | |
BC002728.2 | - | 6 - 7 | 1003 | 64 | |
BC008851.2 | - | 6 - 7 | 1225 | 64 | |
BC035137.1 | - | 6 - 7 | 1163 | 64 | |
CR541915.1 | - | 5 - 6 | 579 | 64 | |
HQ692859.1 | - | 5 - 6 | 579 | 64 | |
J03239.1 | - | 5 - 6 | 636 | 64 | |
M24899.1 | - | 6 - 7 | 990 | 64 | |
X55005.1 | - | 6 - 7 | 1045 | 64 | |
Y00479.1 | - | 6 - 7 | 912 | 64 |