The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000383.3 | NP_000374.1 | 7 - 8 | 1047 | 62 |
GenBank mRNA | AB006682.1 | - | 7 - 8 | 1005 | 62 |
Z97990.1 | - | 7 - 8 | 998 | 62 |