The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000383.3 | NP_000374.1 | 8 - 9 | 1167 | 157 |
GenBank mRNA | AB006682.1 | - | 8 - 9 | 1125 | 157 |
AB006683.1 | - | 2 - 3 | 643 | 157 | |
AB006685.1 | - | 2 - 3 | 643 | 157 | |
AK309572.1 | - | 8 - 9 | 1729 | 157 | |
BC137268.1 | - | 1 - 2 | 824 | 157 | |
BC137270.1 | - | 1 - 2 | 823 | 157 | |
Z97990.1 | - | 8 - 9 | 1118 | 157 |