The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000859.2 | NP_000850.1 | 12 - 13 | 1713 | 94 |
XM_011543357.1 | XP_011541659.1 | 12 - 13 | 1864 | 94 | |
XM_011543358.1 | XP_011541660.1 | 12 - 13 | 1751 | 94 | |
GenBank mRNA | AK292892.1 | - | 12 - 13 | 1644 | 94 |
AK296499.1 | - | 12 - 13 | 1868 | 94 | |
AK312437.1 | - | 11 - 12 | 1557 | 94 | |
M11058.1 | - | 12 - 13 | 1607 | 94 |