The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000859.2 | NP_000850.1 | 14 - 15 | 2030 | 92 |
NM_001130996.1 | NP_001124468.1 | 13 - 14 | 1871 | 92 | |
XM_011543357.1 | XP_011541659.1 | 14 - 15 | 2181 | 92 | |
XM_011543358.1 | XP_011541660.1 | 14 - 15 | 2068 | 92 | |
XM_011543359.1 | XP_011541661.1 | 13 - 14 | 2022 | 92 | |
GenBank mRNA | AK292892.1 | - | 14 - 15 | 1961 | 92 |
AK296499.1 | - | 14 - 15 | 2185 | 92 | |
AK312437.1 | - | 13 - 14 | 1874 | 92 | |
AY429541.1 | - | 2 - 3 | 257 | 92 | |
BC033692.1 | - | 13 - 14 | 1807 | 92 | |
M11058.1 | - | 14 - 15 | 1924 | 92 |