Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL8A
Entrez Gene ID
127829
Gene Name
ADP ribosylation factor like GTPase 8A
Gene Aliases
ARL10B, GIE2
Chromosome Location
Chr.1: 202133404 - 202144743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138795.3 NP_620150.1 4 - 5 543 73
GenBank mRNA AB118752.1 - 4 - 5 372 73
AK127138.1 - 4 - 5 496 73
AK313122.1 - 4 - 5 521 73
BC015408.1 - 4 - 5 474 73
BM548962.1 - 4 - 5 528 73

View Full Product Details