Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITGB4
Entrez Gene ID
3691
Gene Name
integrin subunit beta 4
Gene Aliases
CD104, GP150
Chromosome Location
Chr.17: 75721328 - 75757819 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000213.4 NP_000204.3 10 - 11 1509 107
NM_001005619.1 NP_001005619.1 9 - 10 1222 107
NM_001005731.2 NP_001005731.1 10 - 11 1509 107
NM_001321123.1 NP_001308052.1 10 - 11 1460 107
XM_005257309.2 XP_005257366.1 10 - 11 1279 107
XM_005257311.4 XP_005257368.1 10 - 11 1498 107
XM_006721866.3 XP_006721929.1 10 - 11 1425 107
XM_006721867.3 XP_006721930.1 10 - 11 1422 107
XM_006721868.3 XP_006721931.1 10 - 11 1419 107
XM_006721870.3 XP_006721933.1 10 - 11 1415 107
XM_011524751.2 XP_011523053.1 10 - 11 1423 107
GenBank mRNA AB208913.1 - 10 - 11 1311 107
AF011375.1 - 9 - 10 1214 107
AK310952.1 - 10 - 11 1279 107
BC109238.1 - 6 - 7 967 107
BC118916.1 - 10 - 11 1271 107
BC126411.1 - 10 - 11 1330 107
BC143738.1 - 10 - 11 1330 107
BC143742.1 - 10 - 11 1281 107
X51841.1 - 10 - 11 1340 107
X52186.1 - 9 - 10 1223 107
X53587.1 - 10 - 11 1401 107

View Full Product Details