The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000213.4 | NP_000204.3 | 22 - 23 | 2908 | 94 |
NM_001005619.1 | NP_001005619.1 | 21 - 22 | 2621 | 94 | |
NM_001005731.2 | NP_001005731.1 | 22 - 23 | 2908 | 94 | |
NM_001321123.1 | NP_001308052.1 | 22 - 23 | 2859 | 94 | |
XM_005257309.2 | XP_005257366.1 | 22 - 23 | 2678 | 94 | |
XM_005257311.4 | XP_005257368.1 | 22 - 23 | 2897 | 94 | |
XM_006721866.3 | XP_006721929.1 | 22 - 23 | 2824 | 94 | |
XM_006721867.3 | XP_006721930.1 | 22 - 23 | 2821 | 94 | |
XM_006721868.3 | XP_006721931.1 | 22 - 23 | 2818 | 94 | |
XM_006721870.3 | XP_006721933.1 | 22 - 23 | 2814 | 94 | |
XM_011524751.2 | XP_011523053.1 | 22 - 23 | 2822 | 94 | |
XM_011524752.2 | XP_011523054.1 | 7 - 8 | 1107 | 94 | |
GenBank mRNA | AB208913.1 | - | 22 - 23 | 2710 | 94 |
AF011375.1 | - | 21 - 22 | 2650 | 94 | |
BC118916.1 | - | 22 - 23 | 2670 | 94 | |
BC126411.1 | - | 22 - 23 | 2729 | 94 | |
BC143738.1 | - | 22 - 23 | 2729 | 94 | |
BC143742.1 | - | 22 - 23 | 2680 | 94 | |
X51841.1 | - | 22 - 23 | 2739 | 94 | |
X52186.1 | - | 21 - 22 | 2622 | 94 | |
X53587.1 | - | 22 - 23 | 2800 | 94 |