The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000213.4 | NP_000204.3 | 29 - 30 | 3954 | 96 |
NM_001005619.1 | NP_001005619.1 | 28 - 29 | 3667 | 96 | |
NM_001005731.2 | NP_001005731.1 | 29 - 30 | 3954 | 96 | |
NM_001321123.1 | NP_001308052.1 | 29 - 30 | 3905 | 96 | |
XM_005257309.2 | XP_005257366.1 | 29 - 30 | 3724 | 96 | |
XM_005257311.4 | XP_005257368.1 | 29 - 30 | 3943 | 96 | |
XM_006721866.3 | XP_006721929.1 | 29 - 30 | 3870 | 96 | |
XM_006721867.3 | XP_006721930.1 | 29 - 30 | 3867 | 96 | |
XM_006721868.3 | XP_006721931.1 | 29 - 30 | 3864 | 96 | |
XM_006721870.3 | XP_006721933.1 | 29 - 30 | 3860 | 96 | |
XM_011524751.2 | XP_011523053.1 | 29 - 30 | 3868 | 96 | |
XM_011524752.2 | XP_011523054.1 | 14 - 15 | 2153 | 96 | |
GenBank mRNA | AB208913.1 | - | 29 - 30 | 3756 | 96 |
BC118916.1 | - | 29 - 30 | 3716 | 96 | |
BC126411.1 | - | 29 - 30 | 3775 | 96 | |
BC143738.1 | - | 29 - 30 | 3775 | 96 | |
BC143742.1 | - | 29 - 30 | 3726 | 96 | |
X51841.1 | - | 29 - 30 | 3785 | 96 | |
X52186.1 | - | 28 - 29 | 3668 | 96 | |
X53587.1 | - | 29 - 30 | 3846 | 96 |