Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP19
Entrez Gene ID
10869
Gene Name
ubiquitin specific peptidase 19
Gene Aliases
ZMYND9
Chromosome Location
Chr.3: 49108046 - 49120938 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199160.1 NP_001186089.1 16 - 17 2712 96
NM_001199161.1 NP_001186090.1 16 - 17 2718 96
NM_001199162.1 NP_001186091.1 16 - 17 2682 96
NM_006677.2 NP_006668.1 15 - 16 2409 96
XM_005264823.2 XP_005264880.1 16 - 17 2719 96
XM_005264824.2 XP_005264881.1 16 - 17 2738 96
XM_005264825.2 XP_005264882.1 16 - 17 2713 96
XM_005264826.2 XP_005264883.1 16 - 17 2713 96
XM_005264827.2 XP_005264884.1 16 - 17 2674 96
XM_005264829.2 XP_005264886.1 16 - 17 2719 96
XM_005264830.2 XP_005264887.1 15 - 16 2416 96
XM_005264831.2 XP_005264888.1 15 - 16 2365 96
XM_006712946.2 XP_006713009.1 16 - 17 2716 96
XM_006712947.2 XP_006713010.1 16 - 17 2665 96
XM_006712948.2 XP_006713011.1 16 - 17 2707 96
XM_006712949.2 XP_006713012.1 16 - 17 2569 96
XM_006712950.2 XP_006713013.1 16 - 17 2668 96
XM_006712951.2 XP_006713014.1 16 - 17 2665 96
XM_006712952.2 XP_006713015.1 16 - 17 2662 96
XM_006712953.1 XP_006713016.1 16 - 17 2633 96
XM_017005615.1 XP_016861104.1 16 - 17 2710 96
XM_017005616.1 XP_016861105.1 16 - 17 2704 96
XM_017005617.1 XP_016861106.1 16 - 17 2668 96
XM_017005618.1 XP_016861107.1 16 - 17 2713 96
XM_017005619.1 XP_016861108.1 16 - 17 2710 96
XM_017005620.1 XP_016861109.1 16 - 17 2704 96
XM_017005621.1 XP_016861110.1 16 - 17 2563 96
XM_017005622.1 XP_016861111.1 16 - 17 2674 96
XM_017005623.1 XP_016861112.1 16 - 17 2557 96
XM_017005624.1 XP_016861113.1 16 - 17 2557 96
XM_017005625.1 XP_016861114.1 15 - 16 2410 96
XM_017005626.1 XP_016861115.1 15 - 16 2423 96
XM_017005627.1 XP_016861116.1 15 - 16 2371 96
XM_017005628.1 XP_016861117.1 15 - 16 2365 96
XM_017005629.1 XP_016861118.1 15 - 16 2480 96
XM_017005630.1 XP_016861119.1 15 - 16 2416 96
XM_017005631.1 XP_016861120.1 15 - 16 2410 96
XM_017005632.1 XP_016861121.1 15 - 16 2410 96
XM_017005633.1 XP_016861122.1 15 - 16 2404 96
XM_017005634.1 XP_016861123.1 15 - 16 2371 96
XM_017005635.1 XP_016861124.1 9 - 10 1147 96
GenBank mRNA AB020698.1 - 15 - 16 2251 96
AK294756.1 - 16 - 17 2605 96
AK300425.1 - 16 - 17 2536 96
BC014934.1 - 11 - 12 1690 96
BC065909.1 - 4 - 5 489 96
BC082241.1 - 13 - 14 1785 96
BC142660.1 - 16 - 17 2588 96
BC142727.1 - 16 - 17 2594 96
BC146752.1 - 15 - 16 2251 96

View Full Product Details