Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP19
Entrez Gene ID
10869
Gene Name
ubiquitin specific peptidase 19
Gene Aliases
ZMYND9
Chromosome Location
Chr.3: 49108046 - 49120938 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199160.1 NP_001186089.1 21 - 22 3536 145
NM_001199161.1 NP_001186090.1 21 - 22 3542 145
NM_001199162.1 NP_001186091.1 21 - 22 3506 145
NM_006677.2 NP_006668.1 20 - 21 3233 145
XM_005264823.2 XP_005264880.1 21 - 22 3543 145
XM_005264824.2 XP_005264881.1 21 - 22 3562 145
XM_005264825.2 XP_005264882.1 21 - 22 3537 145
XM_005264826.2 XP_005264883.1 21 - 22 3537 145
XM_005264827.2 XP_005264884.1 21 - 22 3498 145
XM_005264829.2 XP_005264886.1 21 - 22 3543 145
XM_005264830.2 XP_005264887.1 20 - 21 3240 145
XM_005264831.2 XP_005264888.1 20 - 21 3189 145
XM_006712946.2 XP_006713009.1 21 - 22 3540 145
XM_006712947.2 XP_006713010.1 21 - 22 3489 145
XM_006712948.2 XP_006713011.1 21 - 22 3531 145
XM_006712949.2 XP_006713012.1 21 - 22 3393 145
XM_006712950.2 XP_006713013.1 21 - 22 3492 145
XM_006712951.2 XP_006713014.1 21 - 22 3489 145
XM_006712952.2 XP_006713015.1 21 - 22 3486 145
XM_006712953.1 XP_006713016.1 21 - 22 3457 145
XM_017005615.1 XP_016861104.1 21 - 22 3534 145
XM_017005616.1 XP_016861105.1 21 - 22 3528 145
XM_017005617.1 XP_016861106.1 21 - 22 3492 145
XM_017005618.1 XP_016861107.1 21 - 22 3537 145
XM_017005619.1 XP_016861108.1 21 - 22 3534 145
XM_017005620.1 XP_016861109.1 21 - 22 3528 145
XM_017005621.1 XP_016861110.1 21 - 22 3387 145
XM_017005622.1 XP_016861111.1 21 - 22 3498 145
XM_017005623.1 XP_016861112.1 21 - 22 3381 145
XM_017005624.1 XP_016861113.1 21 - 22 3381 145
XM_017005625.1 XP_016861114.1 20 - 21 3234 145
XM_017005626.1 XP_016861115.1 20 - 21 3247 145
XM_017005627.1 XP_016861116.1 20 - 21 3195 145
XM_017005628.1 XP_016861117.1 20 - 21 3189 145
XM_017005629.1 XP_016861118.1 20 - 21 3304 145
XM_017005630.1 XP_016861119.1 20 - 21 3240 145
XM_017005631.1 XP_016861120.1 20 - 21 3234 145
XM_017005632.1 XP_016861121.1 20 - 21 3234 145
XM_017005633.1 XP_016861122.1 20 - 21 3228 145
XM_017005634.1 XP_016861123.1 20 - 21 3195 145
XM_017005635.1 XP_016861124.1 14 - 15 1971 145
GenBank mRNA AB020698.1 - 20 - 21 3075 145
AK294756.1 - 21 - 22 3429 145
AK300425.1 - 21 - 22 3360 145
BC014934.1 - 16 - 17 2514 145
BC048269.1 - 2 - 3 381 145
BC065909.1 - 9 - 10 1313 145
BC082241.1 - 18 - 19 2609 145
BC142660.1 - 21 - 22 3412 145
BC142727.1 - 21 - 22 3418 145
BC146752.1 - 20 - 21 3075 145

View Full Product Details