The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114172.1 | NP_001107644.1 | 8 - 9 | 1202 | 96 |
NM_001303427.1 | NP_001290356.1 | 7 - 8 | 1107 | 96 | |
NM_001303428.1 | NP_001290357.1 | 8 - 9 | 1046 | 96 | |
NM_003629.3 | NP_003620.3 | 7 - 8 | 1701 | 96 | |
XM_017002614.1 | XP_016858103.1 | 8 - 9 | 1079 | 96 | |
GenBank mRNA | AK302049.1 | - | 7 - 8 | 1100 | 96 |
AK304713.1 | - | 2 - 3 | 391 | 96 | |
AK313726.1 | - | 8 - 9 | 1214 | 96 | |
BC021622.1 | - | 7 - 8 | 1635 | 96 | |
CR749649.1 | - | 7 - 8 | 1622 | 96 | |
D88532.1 | - | 7 - 8 | 1365 | 96 |