Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RTN4
- Entrez Gene ID
-
57142
- Gene Name
- reticulon 4
- Gene Aliases
-
ASY, NI220/250, NOGO, NOGO-A, NOGOC, NSP, NSP-CL, Nbla00271, Nbla10545, Nogo-B, Nogo-C, RTN-X, RTN4-A, RTN4-B1, RTN4-B2, RTN4-C
- Chromosome Location
-
Chr.2: 54972187 - 55137831 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB015639.1 |
- |
7 - 8 |
1651 |
108 |
View Full Product Details