Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUP85
Entrez Gene ID
79902
Gene Name
nucleoporin 85
Gene Aliases
FROUNT, Nup75
Chromosome Location
Chr.17: 75205450 - 75235759 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303276.1 NP_001290205.1 13 - 14 1618 59
NM_024844.4 NP_079120.1 14 - 15 1712 59
XM_005257692.3 XP_005257749.1 12 - 13 1301 59
XM_006722094.1 XP_006722157.1 13 - 14 1337 59
XM_011525267.1 XP_011523569.1 13 - 14 1350 59
XM_017025108.1 XP_016880597.1 12 - 13 1515 59
XM_017025109.1 XP_016880598.1 12 - 13 1376 59
XM_017025110.1 XP_016880599.1 12 - 13 1242 59
GenBank mRNA AF498261.1 - 14 - 15 1400 59
AF514995.1 - 14 - 15 1480 59
AK022611.1 - 14 - 15 1442 59
AK124501.1 - 4 - 5 1277 59
AK297574.1 - 13 - 14 1361 59
AK298519.1 - 13 - 14 1320 59
AL833893.1 - 14 - 15 1405 59
BC000697.2 - 14 - 15 1471 59

View Full Product Details