The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146705.1 | NP_001140177.1 | 21 - 22 | 3324 | 80 |
NM_001146706.1 | NP_001140178.1 | 19 - 20 | 3060 | 80 | |
NM_004653.4 | NP_004644.2 | 20 - 21 | 3231 | 80 | |
XM_005262560.2 | XP_005262617.1 | 19 - 20 | 3504 | 80 | |
XM_005262561.2 | XP_005262618.1 | 20 - 21 | 3408 | 80 | |
XM_011531468.2 | XP_011529770.1 | 20 - 21 | 3639 | 80 | |
GenBank mRNA | AK095923.1 | - | 4 - 5 | 436 | 80 |
AK127269.1 | - | 9 - 10 | 2995 | 80 | |
BC132721.1 | - | 20 - 21 | 3154 | 80 | |
BC144101.1 | - | 21 - 22 | 3247 | 80 | |
BC144102.1 | - | 21 - 22 | 3247 | 80 | |
BC146767.1 | - | 19 - 20 | 2882 | 80 | |
D87072.1 | - | 19 - 20 | 2882 | 80 | |
U52191.1 | - | 20 - 21 | 3218 | 80 |